Assay Design
Gene Transcripts
- Gene Symbol
- ACA4
- Entrez Gene ID
-
827846
- Gene Name
- alpha carbonic anhydrase 4
- Gene Aliases
-
A. THALIANA ALPHA CARBONIC ANHYDRASE 4, AT4G20990, ATACA4, T13K14.1, T13K14_1, alpha carbonic anhydrase 4
- Chromosome Location
-
Chr.4: 11219772 - 11221148 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118217.2 |
NP_193831.1 |
2 - 3 |
306 |
78 |
View Full Product Details