Assay Design
Gene Transcripts
- Gene Symbol
- HSA32
- Entrez Gene ID
-
827880
- Gene Name
- Aldolase-type TIM barrel family protein
- Gene Aliases
-
AT4G21320, HEAT-STRESS-ASSOCIATED 32, T6K22.50, T6K22_50
- Chromosome Location
-
Chr.4: 11340362 - 11342075 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118250.5 |
NP_567623.2 |
3 - 4 |
656 |
93 |
GenBank mRNA |
AK118775.1 |
- |
3 - 4 |
569 |
93 |
View Full Product Details