Assay Design
Gene Transcripts
- Gene Symbol
- CPL1
- Entrez Gene ID
-
828254
- Gene Name
- C-terminal domain phosphatase-like 1
- Gene Aliases
-
AT4G21670, ATCPL1, ATCPL2, C-TERMINAL DOMAIN PHOSPHATASE-LIKE 1, C-terminal domain phosphatase-like 1, F17L22.130, F17L22_130, FIERY 2, FRY2, RCF2, SHI4, SHINY 4, regulators of C-REPEAT BINDING FACTOR (CBF) gene expression 2
- Chromosome Location
-
Chr.4: 11510989 - 11517396 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118287.5 |
NP_193898.3 |
12 - 13 |
2893 |
65 |
GenBank mRNA |
AY557186.1 |
- |
12 - 13 |
2371 |
65 |
View Full Product Details