Assay Design



Gene Transcripts

Gene Symbol
BGLU47
Entrez Gene ID
828264
Gene Name
beta-glucosidase 47
Gene Aliases
AT4G21760, F17L22.220, F17L22_220, beta-glucosidase 47
Chromosome Location
Chr.4: 11561123 - 11563950 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341512.1 NP_001328261.1 1 - 2 328 64
NM_118296.4 NP_193907.2 1 - 2 250 64

View Full Product Details