Assay Design



Gene Transcripts

Gene Symbol
CPK15
Entrez Gene ID
828283
Gene Name
calcium-dependent protein kinase 15
Gene Aliases
AT4G21940, F1N20.5, calcium-dependent protein kinase 15
Chromosome Location
Chr.4: 11640735 - 11643762 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203865.1 NP_001190794.1 2 - 3 952 148
NM_001341523.1 NP_001329845.1 2 - 3 1019 148
NM_118315.2 NP_193925.1 2 - 3 1019 148
GenBank mRNA AY034995.1 - 2 - 3 947 148
AY063004.1 - 2 - 3 907 148

View Full Product Details