Assay Design



Gene Transcripts

Gene Symbol
CPK15
Entrez Gene ID
828283
Gene Name
calcium-dependent protein kinase 15
Gene Aliases
AT4G21940, F1N20.5, calcium-dependent protein kinase 15
Chromosome Location
Chr.4: 11640735 - 11643762 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203865.1 NP_001190794.1 3 - 4 1112 67
NM_001341523.1 NP_001329845.1 3 - 4 1179 67
NM_118315.2 NP_193925.1 3 - 4 1179 67
GenBank mRNA AY034995.1 - 3 - 4 1107 67
AY063004.1 - 3 - 4 1067 67

View Full Product Details