Assay Design



Gene Transcripts

Gene Symbol
sks4
Entrez Gene ID
828290
Gene Name
SKU5 similar 4
Gene Aliases
AT4G22010, F1N20.110, F1N20_110, SKU5 similar 4, SKU5 similar 4
Chromosome Location
Chr.4: 11663186 - 11666716 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118322.4 NP_193932.1 2 - 3 491 122
GenBank mRNA AY081335.1 - 2 - 3 353 122
BT008899.1 - 2 - 3 248 122

View Full Product Details