Assay Design
Gene Transcripts
- Gene Symbol
- AT4G22755
- Entrez Gene ID
-
28720173
- Gene Name
- methylsterol monooxygenase 1-3
- Gene Aliases
-
AT4G22755
- Chromosome Location
-
Chr.4: 11951830 - 11953731 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118403.2 |
NP_567669.1 |
1 - 2 |
451 |
91 |
View Full Product Details