Assay Design



Gene Transcripts

Gene Symbol
CRK10
Entrez Gene ID
828417
Gene Name
cysteine-rich RLK (RECEPTOR-like protein kinase) 10
Gene Aliases
AT4G23180, F21P8.70, F21P8_70, RLK4, cysteine-rich RLK (RECEPTOR-like protein kinase) 10
Chromosome Location
Chr.4: 12137995 - 12140930 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118447.3 NP_567679.2 6 - 7 1865 61
GenBank mRNA AK117765.1 - 6 - 7 1723 61
BT005954.1 - 6 - 7 1689 61

View Full Product Details