Assay Design
Gene Transcripts
- Gene Symbol
- WRKY29
- Entrez Gene ID
-
828455
- Gene Name
- WRKY family transcription factor
- Gene Aliases
-
AT4G23550, ATWRKY29, F9D16.20, F9D16_20
- Chromosome Location
-
Chr.4: 12291632 - 12293367 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118486.6 |
NP_194086.4 |
1 - 2 |
541 |
125 |
View Full Product Details