Assay Design



Gene Transcripts

Gene Symbol
TRH1
Entrez Gene ID
828464
Gene Name
Potassium transporter family protein
Gene Aliases
AT4G23640, ATKT3, F9D16.110, F9D16_110, KUP4, TINY ROOT HAIR 1
Chromosome Location
Chr.4: 12320152 - 12324604 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118495.4 NP_194095.2 7 - 8 1293 120
GenBank mRNA AJ296155.1 - 7 - 8 1131 120
AK226593.1 - 7 - 8 1161 120

View Full Product Details