Assay Design



Gene Transcripts

Gene Symbol
UPS2
Entrez Gene ID
814882
Gene Name
ureide permease 2
Gene Aliases
ARABIDOPSIS THALIANA UREIDE PERMEASE 2, AT2G03530, ATUPS2, T4M8.3, T4M8_3, ureide permease 2
Chromosome Location
Chr.2: 1071453 - 1074016 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084405.2 NP_001077874.1 1 - 2 491 69
NM_001335189.1 NP_001324119.1 1 - 2 673 69
NM_126403.4 NP_178451.2 2 - 3 499 69

View Full Product Details