Assay Design



Gene Transcripts

Gene Symbol
SEP4
Entrez Gene ID
814898
Gene Name
K-box region and MADS-box transcription factor family protein
Gene Aliases
AGAMOUS-like 3, AGL3, AT2G03710, F19B11.16, F19B11_16, MADS BOX PROTEIN, SEPALLATA 4
Chromosome Location
Chr.2: 1129229 - 1131838 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126418.2 NP_178466.1 3 - 4 677 127
NM_179599.2 NP_849930.1 3 - 4 716 127
NM_201682.3 NP_973411.1 3 - 4 711 127
GenBank mRNA AY063894.1 - 3 - 4 656 127
AY096386.1 - 3 - 4 323 127
AY141229.1 - 3 - 4 323 127
U81369.1 - 3 - 4 677 127

View Full Product Details