Assay Design



Gene Transcripts

Gene Symbol
VPS25
Entrez Gene ID
827637
Gene Name
E2F/DP family winged-helix DNA-binding domain-containing protein
Gene Aliases
AT4G19003
Chromosome Location
Chr.4: 10406694 - 10409057 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341313.1 NP_001319991.1 5 - 6 499 101
NM_001341314.1 NP_001319990.1 5 - 6 712 101
NM_001341315.1 NP_001329781.1 5 - 6 499 101
NM_001341316.1 NP_001329780.1 5 - 6 618 101
NM_148354.4 NP_680720.1 5 - 6 454 101
NM_202841.2 NP_974570.1 5 - 6 618 101
GenBank mRNA AK317105.1 - 5 - 6 601 101
AY065055.1 - 5 - 6 454 101
AY093752.1 - 4 - 5 343 101

View Full Product Details