Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001341501.1 | NP_001328108.1 | 4 - 5 | 794 | 126 |
NM_001341502.1 | NP_001328109.1 | 4 - 5 | 766 | 126 | |
NM_001341503.1 | NP_001328107.1 | 6 - 7 | 576 | 126 | |
NM_001341504.1 | NP_001328110.1 | 4 - 5 | 746 | 126 | |
NM_148368.2 | NP_680734.1 | 5 - 6 | 716 | 126 |