Assay Design
Gene Transcripts
- Gene Symbol
- PP2-A10
- Entrez Gene ID
-
837553
- Gene Name
- phloem protein 2-A10
- Gene Aliases
-
AT1G10155, ATPP2-A10, phloem protein 2-A10
- Chromosome Location
-
Chr.1: 3325825 - 3328448 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_148455.2 |
NP_683296.1 |
1 - 2 |
58 |
81 |
View Full Product Details