Assay Design



Gene Transcripts

Gene Symbol
NUDX25
Entrez Gene ID
839890
Gene Name
nudix hydrolase homolog 25
Gene Aliases
AT1G30110, ATNUDX25, T2H7.9, T2H7_9, nudix hydrolase homolog 25
Chromosome Location
Chr.1: 10581735 - 10584152 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198185.1 NP_001185114.1 5 - 6 403 108
NM_102750.5 NP_174303.1 4 - 5 355 108
GenBank mRNA BT025530.1 - 3 - 4 212 108

View Full Product Details