Assay Design



Gene Transcripts

Gene Symbol
ABCC11
Entrez Gene ID
839922
Gene Name
multidrug resistance-associated protein 12
Gene Aliases
AT1G30420, ATMRP12, ATP-binding cassette C11, MRP12, T4K22.1, T4K22_1, multidrug resistance-associated protein 12
Chromosome Location
Chr.1: 10748261 - 10756695 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332898.1 NP_001319112.1 25 - 26 4290 87
NM_001332899.1 NP_001321245.1 23 - 24 4452 87

View Full Product Details