Assay Design



Gene Transcripts

Gene Symbol
PHV
Entrez Gene ID
839928
Gene Name
Homeobox-leucine zipper family protein / lipid-binding START domain-containing protein
Gene Aliases
AT1G30490, ATHB9, F26G16.11, F26G16_11, PHAVOLUTA
Chromosome Location
Chr.1: 10796124 - 10801093 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102785.5 NP_174337.1 10 - 11 1570 82
GenBank mRNA AJ440967.1 - 10 - 11 1221 82
AK226570.1 - 10 - 11 1435 82

View Full Product Details