Assay Design



Gene Transcripts

Gene Symbol
PRR1
Entrez Gene ID
840102
Gene Name
pinoresinol reductase 1
Gene Aliases
AT1G32100, ATPRR1, F3C3.10, F3C3_10, PSEUDO-RESPONSE REGULATOR 1, pinoresinol reductase 1
Chromosome Location
Chr.1: 11546160 - 11548016 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102944.3 NP_174490.1 3 - 4 856 64
GenBank mRNA AY065214.1 - 3 - 4 834 64
AY096520.1 - 3 - 4 793 64

View Full Product Details