Assay Design
Gene Transcripts
- Gene Symbol
- PLC1
- Entrez Gene ID
-
830010
- Gene Name
- phospholipase C1
- Gene Aliases
-
AT4G38530, ATPLC1, ATPLC3, PLC3, Phosphoinositide-Specific Phospholipase C Isoform 3, phospholipase C1
- Chromosome Location
-
Chr.4: 18020567 - 18022985 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_120014.5 |
NP_195565.2 |
4 - 5 |
1167 |
103 |
GenBank mRNA |
AK222239.1 |
- |
4 - 5 |
1060 |
103 |
View Full Product Details