Assay Design



Gene Transcripts

Gene Symbol
PIS2
Entrez Gene ID
830014
Gene Name
Putative CDP-diacylglycerol-inositol 3-phosphatidyltransferase 2
Gene Aliases
AT4G38570, F20M13.130, F20M13_130, probable CDP-diacylglycerol--inositol 3-phosphatidyltransferase 2
Chromosome Location
Chr.4: 18031279 - 18033599 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204026.2 NP_001190955.1 4 - 5 502 93
NM_001342505.1 NP_001328584.1 3 - 4 697 93
NM_001342506.1 NP_001328583.1 4 - 5 544 93
NM_120018.4 NP_195569.1 4 - 5 500 93
GenBank mRNA AY086801.1 - 4 - 5 400 93

View Full Product Details