Assay Design



Gene Transcripts

Gene Symbol
BPC5
Entrez Gene ID
830046
Gene Name
basic pentacysteine 5
Gene Aliases
AT4G38910, ATBPC5, BBR/BPC5, F19H22.10, F19H22_10, basic pentacysteine 5
Chromosome Location
Chr.4: 18145188 - 18147080 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204028.2 NP_001190957.1 2 - 3 177 70

View Full Product Details