Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001342527.1 | NP_001328715.1 | 11 - 12 | 1233 | 66 |
NM_120081.2 | NP_195632.1 | 11 - 12 | 1411 | 66 | |
GenBank mRNA | AY059862.1 | - | 11 - 12 | 1351 | 66 |
BT010378.1 | - | 10 - 11 | 1192 | 66 |