Assay Design



Gene Transcripts

Gene Symbol
LIP1
Entrez Gene ID
816012
Gene Name
lipase 1
Gene Aliases
AT2G15230, ATLIP1, F15A23.3, F15A23_3, lipase 1
Chromosome Location
Chr.2: 6612480 - 6615457 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127084.3 NP_179126.2 6 - 7 876 95
GenBank mRNA AF530477.1 - 6 - 7 717 95
AK229578.1 - 6 - 7 775 95
BT025547.1 - 6 - 7 690 95

View Full Product Details