Assay Design
Gene Transcripts
- Gene Symbol
- SSP
- Entrez Gene ID
-
816214
- Gene Name
- kinase with tetratricopeptide repeat domain-containing protein
- Gene Aliases
-
AT2G17090, BSK12, F6P23.23, F6P23_23, SHORT SUSPENSOR, brassinosteroid-signaling kinase 12
- Chromosome Location
-
Chr.2: 7434919 - 7437314 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_127264.2 |
NP_179301.1 |
5 - 6 |
712 |
116 |
View Full Product Details