Assay Design



Gene Transcripts

Gene Symbol
PHT3;3
Entrez Gene ID
816233
Gene Name
phosphate transporter 3;3
Gene Aliases
AT2G17270, F5J6.3, F5J6_3, MPT1, mitochondrial phosphate transporter 1, phosphate transporter 3;3
Chromosome Location
Chr.2: 7510097 - 7512502 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127282.3 NP_179319.1 4 - 5 1111 97
GenBank mRNA AK227300.1 - 4 - 5 1109 97
BT010199.1 - 4 - 5 752 97

View Full Product Details