Assay Design
Gene Transcripts
- Gene Symbol
- NIP1
- Entrez Gene ID
-
816284
- Gene Name
- NEP-interacting protein 1
- Gene Aliases
-
AT2G17750, NEP-interacting protein 1, T17A5.3, T17A5_3
- Chromosome Location
-
Chr.2: 7709192 - 7711151 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_127329.2 |
NP_179366.1 |
3 - 4 |
461 |
161 |
GenBank mRNA |
DQ446513.1 |
- |
3 - 4 |
461 |
161 |
View Full Product Details