Assay Design
Gene Transcripts
- Gene Symbol
- HK1
- Entrez Gene ID
-
816291
- Gene Name
- histidine kinase 1
- Gene Aliases
-
AHK1, AT2G17820, ATHK1, T13L16.16, T13L16_16, histidine kinase 1
- Chromosome Location
-
Chr.2: 7742870 - 7748502 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_127335.2 |
NP_565424.1 |
1 - 2 |
737 |
127 |
View Full Product Details