Assay Design
Gene Transcripts
- Gene Symbol
- CPK16
- Entrez Gene ID
-
816299
- Gene Name
- calcium-dependent protein kinase 16
- Gene Aliases
-
AT2G17890, AtCDPK16, CDPK16, T13L16.9, T13L16_9, calcium-dependent protein kinase 16
- Chromosome Location
-
Chr.2: 7769521 - 7772782 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_127343.2 |
NP_179379.1 |
5 - 6 |
945 |
92 |
View Full Product Details