Assay Design



Gene Transcripts

Gene Symbol
PAP11
Entrez Gene ID
816326
Gene Name
purple acid phosphatase 11
Gene Aliases
AT2G18130, ATPAP11, F8D23.9, F8D23_9, PURPLE ACID PHOSPHATASE 11, purple acid phosphatase 11
Chromosome Location
Chr.2: 7879308 - 7883935 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127370.1 NP_179405.1 5 - 6 791 78
GenBank mRNA AF492663.1 - 5 - 6 791 78
BT026132.1 - 5 - 6 791 78

View Full Product Details