Assay Design
Gene Transcripts
- Gene Symbol
- SHP2
- Entrez Gene ID
-
818883
- Gene Name
- K-box region and MADS-box transcription factor family protein
- Gene Aliases
-
AGAMOUS-like 5, AGL5, AT2G42830, F7D19.17, F7D19_17, SHATTERPROOF 2
- Chromosome Location
-
Chr.2: 17820114 - 17824013 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_180046.2 |
NP_850377.1 |
4 - 5 |
658 |
153 |
GenBank mRNA |
BT029453.1 |
- |
3 - 4 |
471 |
153 |
View Full Product Details