Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202847.1 | NP_001189776.1 | 1 - 2 | 235 | 78 |
NM_130374.4 | NP_566122.1 | 1 - 2 | 337 | 78 | |
NM_180153.2 | NP_850484.1 | 1 - 2 | 208 | 78 | |
GenBank mRNA | AY045783.1 | - | 1 - 2 | 202 | 78 |
AY079358.1 | - | 1 - 2 | 189 | 78 |