Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001338031.1 | NP_001327928.1 | 3 - 4 | 3627 | 102 |
NM_112147.3 | NP_187915.1 | 3 - 4 | 3627 | 102 | |
NM_180244.1 | NP_850575.1 | 3 - 4 | 3158 | 102 | |
NM_202570.1 | NP_974299.1 | 3 - 4 | 3158 | 102 | |
NM_202571.1 | NP_974300.1 | 3 - 4 | 3158 | 102 |