Assay Design



Gene Transcripts

Gene Symbol
MSH7
Entrez Gene ID
822040
Gene Name
MUTS homolog 7
Gene Aliases
ARABIDOPSIS THALIANA MUTS HOMOLOG 7, AT3G24495, ATMSH7, MSH6-2, MUTS HOMOLOG 6-2, MUTS homolog 7
Chromosome Location
Chr.3: 8912343 - 8918288 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338685.1 NP_001326470.1 8 - 9 1553 97
NM_180299.1 NP_850630.1 8 - 9 1411 97

View Full Product Details