Assay Design



Gene Transcripts

Gene Symbol
DCL1
Entrez Gene ID
839574
Gene Name
dicer-like 1
Gene Aliases
ABNORMAL SUSPENSOR 1, ASU1, AT1G01040, ATDCL1, CAF, CARPEL FACTORY, DICER-LIKE 1, EMB60, EMB76, EMBRYO DEFECTIVE 60, EMBRYO DEFECTIVE 76, SHORT INTEGUMENTS 1, SIN1, SUS1, SUSPENSOR 1, T25K16.4, T25K16_4, dicer-like 1
Chromosome Location
Chr.1: 23121 - 31227 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197952.1 NP_001184881.1 3 - 4 1363 83
NM_099986.4 NP_171612.1 3 - 4 1658 83
GenBank mRNA AF292940.1 - 3 - 4 1633 83

View Full Product Details