Assay Design



Gene Transcripts

Gene Symbol
KCS1
Entrez Gene ID
839395
Gene Name
3-ketoacyl-CoA synthase 1
Gene Aliases
3-ketoacyl-CoA synthase 1, AT1G01120, T25K16.11, T25K16_11
Chromosome Location
Chr.1: 57164 - 59215 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_099994.4 NP_171620.2 1 - 1 1787 78
GenBank mRNA AY074285.1 - 1 - 1 1739 78
AY096568.1 - 1 - 1 1550 78

View Full Product Details