Assay Design



Gene Transcripts

Gene Symbol
AKN2
Entrez Gene ID
830153
Gene Name
APS-kinase 2
Gene Aliases
ADENOSINE-5'-PHOSPHOSULFATE (APS) KINASE 2, ADENOSINE-5'-PHOSPHOSULFATE-KINASE, APK2, APS-kinase 2, AT4G39940, T5J17.110, T5J17_110
Chromosome Location
Chr.4: 18519599 - 18521605 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120157.4 NP_195704.1 1 - 2 415 87
GenBank mRNA AF043351.2 - 1 - 2 266 87
AF462823.1 - 1 - 2 296 87
AY097421.1 - 1 - 2 227 87

View Full Product Details