Assay Design



Gene Transcripts

Gene Symbol
PTR5
Entrez Gene ID
831725
Gene Name
peptide transporter 5
Gene Aliases
ARABIDOPSIS THALIANA PEPTIDE TRANSPORTER 5, AT5G01180, ATPTR5, AtNPF8.2, F7J8.160, F7J8_160, NPF8.2, NRT1/ PTR family 8.2, peptide transporter 5, peptide transporter 5
Chromosome Location
Chr.5: 61017 - 63936 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342576.1 NP_001331271.1 4 - 5 1107 76
NM_001342577.1 NP_001331272.1 4 - 5 1092 76
NM_120196.4 NP_195738.1 4 - 5 1068 76

View Full Product Details