Assay Design



Gene Transcripts

Gene Symbol
LAX1
Entrez Gene ID
831866
Gene Name
like AUXIN RESISTANT 1
Gene Aliases
AT5G01240, F7J8.220, F7J8_220, like AUXIN RESISTANT 1
Chromosome Location
Chr.5: 97536 - 101835 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120202.5 NP_195744.1 4 - 5 1056 92
NM_202990.2 NP_974719.1 2 - 3 380 92

View Full Product Details