Assay Design



Gene Transcripts

Gene Symbol
CPL2
Entrez Gene ID
831743
Gene Name
carboxyl-terminal domain (ctd) phosphatase-like 2
Gene Aliases
AT5G01270, ATCPL2, F7J8.250, F7J8_250, carboxyl-terminal domain (ctd) phosphatase-like 2
Chromosome Location
Chr.5: 107547 - 112324 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203270.1 NP_001190199.1 13 - 14 2331 57
NM_120205.5 NP_195747.2 13 - 14 2344 57
GenBank mRNA AY557187.1 - 13 - 14 2163 57

View Full Product Details