Assay Design



Gene Transcripts

Gene Symbol
CAX4
Entrez Gene ID
831754
Gene Name
cation exchanger 4
Gene Aliases
AT5G01490, ATCAX4, F7A7.10, F7A7_10, cation exchanger 4
Chromosome Location
Chr.5: 195452 - 198651 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342594.1 NP_001331104.1 6 - 7 1080 67
NM_120227.4 NP_568091.2 6 - 7 1080 67

View Full Product Details