Assay Design
Gene Transcripts
- Gene Symbol
- STN8
- Entrez Gene ID
-
831741
- Gene Name
- Protein kinase superfamily protein
- Gene Aliases
-
AT5G01920, STATE TRANSITION 8, State transition 8, T20L15.190, T20L15_190
- Chromosome Location
-
Chr.5: 359107 - 361310 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_120270.4 |
NP_195812.1 |
4 - 5 |
1747 |
97 |
View Full Product Details