Assay Design



Gene Transcripts

Gene Symbol
RPL
Entrez Gene ID
831745
Gene Name
POX (plant homeobox) family protein
Gene Aliases
AT5G02030, BEL1-LIKE HOMEODOMAIN 9, BELLRINGER, BLH9, BLR, HB-6, LARSON, LSN, PENNYWISE, PNY, REPLUMLESS, T7H20.80, T7H20_80, VAAMANA, VAN
Chromosome Location
Chr.5: 395567 - 399125 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120281.3 NP_195823.1 1 - 2 901 95
GenBank mRNA AY328314.1 - 1 - 2 834 95
BT020537.1 - 1 - 2 741 95

View Full Product Details