Assay Design



Gene Transcripts

Gene Symbol
EER5
Entrez Gene ID
816475
Gene Name
proteasome family protein
Gene Aliases
AT2G19560, AtTHP1, ENHANCED ETHYLENE RESPONSE 5, ESSP1, F3P11.16, F3P11_16, ectopic expression of seed storage proteins 1
Chromosome Location
Chr.2: 8465690 - 8469145 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335638.1 NP_001324728.1 6 - 7 484 73
NM_127514.4 NP_179546.2 8 - 9 836 73
GenBank mRNA AK118897.1 - 8 - 9 807 73

View Full Product Details