Assay Design



Gene Transcripts

Gene Symbol
TET2
Entrez Gene ID
816477
Gene Name
tetraspanin2
Gene Aliases
AT2G19580, F3P11.18, F3P11_18, tetraspanin2
Chromosome Location
Chr.2: 8472134 - 8475153 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127516.4 NP_179548.1 2 - 3 637 72
GenBank mRNA AK229021.1 - 2 - 3 575 72
BT011213.1 - 2 - 3 549 72
BT012532.1 - 2 - 3 505 72

View Full Product Details