Assay Design
Gene Transcripts
- Gene Symbol
- MIOX2
- Entrez Gene ID
-
816499
- Gene Name
- myo-inositol oxygenase 2
- Gene Aliases
-
AT2G19800, F6F22.17, F6F22_17, myo-inositol oxygenase 2
- Chromosome Location
-
Chr.2: 8530865 - 8533779 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_127538.4 |
NP_565459.1 |
8 - 9 |
1194 |
85 |
GenBank mRNA |
BT033135.1 |
- |
8 - 9 |
769 |
85 |
View Full Product Details