Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001335672.1 | NP_001325368.1 | 20 - 21 | 2530 | 72 |
NM_001335673.1 | NP_001325369.1 | 20 - 21 | 2530 | 72 | |
NM_001335674.1 | NP_001325367.1 | 20 - 21 | 2537 | 72 | |
NM_127588.3 | NP_179619.2 | 20 - 21 | 2339 | 72 |