Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_127622.4 | NP_179650.1 | 2 - 3 | 832 | 66 |
NM_201760.2 | NP_973489.1 | 2 - 3 | 780 | 66 | |
GenBank mRNA | AF301898.1 | - | 2 - 3 | 745 | 66 |
AY050987.1 | - | 2 - 3 | 777 | 66 | |
AY091293.1 | - | 2 - 3 | 690 | 66 |