Assay Design



Gene Transcripts

Gene Symbol
SUR1
Entrez Gene ID
816585
Gene Name
Tyrosine transaminase family protein
Gene Aliases
ABERRANT LATERAL ROOT FORMATION 1, ALF1, AT2G20610, F23N11.7, F23N11_7, HLS3, HOOKLESS 3, ROOTY, ROOTY 1, RTY, RTY1, SUPERROOT 1
Chromosome Location
Chr.2: 8877887 - 8880440 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127622.4 NP_179650.1 5 - 6 1261 109
NM_201760.2 NP_973489.1 5 - 6 1209 109
GenBank mRNA AF301898.1 - 5 - 6 1174 109
AY050987.1 - 5 - 6 1206 109
AY091293.1 - 5 - 6 1119 109

View Full Product Details